On the Process and Review page, the Results table displays information about the components identified in your peptide mapping analysis experiment, including identifications, modifications, sequence information, retention times, and more.
Prerequisites
  1. You have opened your peptide mapping analysis results and are viewing the Process and Review page.
Procedure
  1. In the Results table on the Process and Review page, select a component row in the table to populate the other panes on the Process and Review page with information related to the selected component.
  2. The Results table is organized by components at the upper level and by raw data files at the lower level. By default, the table displays results at the component level (top-level data).
  3. If your analysis contains multiple raw data files, you can view results related to a specific raw data file for each component (lower-level data).
  4. Select the plus icon (+) in the left margin of a component row to display the component information at the raw data file level.
Process and Review page for peptide mapping analysis
Process and Review page for peptide mapping analysis
TIP
For more information on the columns and information reported in the Results tables, refer to the BioPharma Finder Software User Guide.