After you submit your peptide mapping analysis for processing, the application opens the Queue page, where you can monitor the status of your experiment. When your experiment is complete, you can open and view the results to identify the appropriate PQAs in your analysis.
Prerequisites
- You have submitted your peptide mapping analysis for processing.
- You are viewing the Queue page.
Procedure
- On the Queue page, in the run queue, do one of the following:
- Double-click the row of a completed job.
- Select the row of a completed job and then select Open Results in the command bar.
- The Process and Review page opens by default.
NOTE
You cannot open experiment results if the application has not completely processed the experiment. The Open Results button is inactive until the selected job is completed. Opening experiment results does not interfere with the application processing subsequent experiments in the run queue.
NOTE
In addition to the Process and Review page, you can also select the Mapping tab to view the Coverage page and Modification Summary page to view additional results. For more information on the results displayed on the Coverage and Modification Summary pages, refer to the BioPharma Finder Software User Guide.