When the MS instrument is running and connected, you have the following options:
  • You can view the instrument queue in the instrument view of the Chromeleon Console, where you can insert your sequence into the queue.
  • You can view the sequence in the data view of the Chromeleon Console, or in the Chromatography Studio, and you can queue your sequence on a selected instrument from the control bar of the sequence.
  • You can manually perform a ready check that verifies that the assigned instrument is ready for operation and that the instrument methods that you used in the queued sequences do not contain errors. After the ready check, you can run the sequence immediately or schedule the run for a later time.
TIP
Refer to the Chromeleon CDS Software User Help for more information on any of the options.