Unlike the BioPharma Finder software, Chromeleon CDS software does not process MS2 spectra. The XICs that you set up are created from full-MS data only; therefore, these chromatograms rely on accurate masses and on reproducible retention times. To confirm the identity of XICs, you must use composite scoring and isotopic distribution charts.