The end goal is to make the attribute monitoring processing method work with data acquired as full-MS scans only. However, you can start refining the processing method even before you have such data acquired with a dedicated instrument method. Chromeleon CDS software does not use data-dependent scans, so you can continue developing the processing method around the data that you acquired for the peptide characterization.