In Chromeleon CDS software, the table of targeted peptides and their data is a part of the processing method. You can have several processing methods saved in a sequence. As a result, you can easily re-process a sequence for different lists of targeted peptides, just by selecting a different processing method.
Prerequisites
- In Chromeleon CDS software, you have created a new processing method. Refer to the Chromeleon CDS Software Quick Start Guide or the Chromeleon CDS Software User Help for the various ways how you can do that.
Procedure
- Open the processing method in the Chromatography Studio.
- If not already selected, select the Data Processing category.
- If not already displayed, open the Processing Method pane in the work area.