A peptide workbook (.wbpf file) contains both a list of the peptides that are selected from the Results table of a peptide mapping analysis experiment and the processing method parameters used to obtain those results. A peptide workbook can contain data from multiple different experiments. For more information about the file format, refer to the BioPharma Finder Software User Guide.
After viewing your peptide mapping results on the Process and Review page, you can select specific PQAs in the Results table to save as a peptide workbook and import into Chromeleon CDS software.
Prerequisites
  1. You have opened your peptide mapping analysis results and are viewing the Process and Review page.
  2. You have filtered the Results table and identified the PQAs of interest.
Procedure
  1. On the Process and Review page, select the checkbox in the row of each PQA in the Results table that you want to save to a workbook.
  2. TIP
    To select all of the rows, select the checkbox in the column header.
  3. Right-click anywhere in the Results table, select Save As Peptide Workbook, and then do one of the following:
  4. Select Save As Peptide Workbook > All to export all results to a workbook.
  5. Select Save As Peptide Workbook > Checked to export only the selected results to a workbook.
  6. NOTE
    The application does not support saving unidentified components or components with unspecified modifications, dimers, adducts, disulfide bonds, or any type of gas phase modifications to the workbook. You must clear the checkboxes for these components to save the workbook.
  7. The Save Peptide Workbook As dialog box opens.
  8. In the Save Peptide Workbook As dialog box, select one of the following options:
  9. Create a New Peptide Workbook
  10. Select an Existing Workbook
  11. (For the Create a New Peptide Workbook option) In the Workbook Name box, enter a new name to create a new workbook or keep the same name to overwrite the workbook if it already exists.
  12. (For the Select an Existing Workbook option) Select the checkbox for an existing workbook in the table.
  13. (Optional) In the Description box, enter a description for the workbook.
  14. In the Number of Isotopes per Peptide box, specify the number of isotopes for the selected components.
  15. Select Save and navigate to the location in the Ardia Platform data repository where you want to save the peptide workbook.
NOTE
After saving, you can manage the workbook using the Target Peptide Workbook page. For more information on managing a target peptide workbook, refer to the BioPharma Finder Software User Guide.