The Thermo Scientific MAM 2.0 workflow includes the following technologies:
Technology | Description |
---|---|
Thermo Scientific™ Vanquish™ Horizon Binary UHPLC System or Thermo Scientific™ Vanquish™ Flex Binary UHPLC System | An ultra-high pressure liquid chromatography platform that provides robust and consistent peptide separations. |
Thermo Scientific™ Orbitrap Exploris™ 240 mass spectrometer or Orbitrap Exploris™ MX mass detector | Mass spectrometry instruments that provide sensitivity and specificity to identify, quantify, and monitor peptides. Orbitrap Exploris 240 is suitable for both the research and development phase and quality and control phase. Orbitrap Exploris MX is suitable for the quality and control phase. |
Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) software | Powerful, compliance-ready software ecosystem for chromatography. |
Thermo Scientific™ BioPharma Finder™ software | Workflow-driven solution for complete characterization of biotherapeutics. |
Thermo Scientific™ Ardia™ Platform | An integrated application and data platform for the seamless sharing and synchronization of data between chromatography and mass spectrometry instruments, applications, and the wider ecosystem. |
NOTE
Different configurations of the workflow are also possible, for example, using the Thermo Scientific™ Orbitrap Exploris™ 480 Mass spectrometer or the Thermo Scientific™ Xcalibur™ software for data acquisition. Such configurations do not qualify as the Thermo Scientific MAM 2.0 workflow that is described in this document.