To run a peptide mapping analysis in the BioPharma Finder software, you must add the protein sequence to the Sequence Manager. This sequence is used to match experimental components in the peptide mapping analysis. Then you set up and run the peptide mapping analysis experiment.