If your Chromeleon data vault is connected to the Ardia Platform, then it is synced with the Ardia Platform after each completed injection. As raw files are acquired, they become automatically available for all other software connected to the Ardia Platform.
For details on connecting your Chromeleon data vault server to the Ardia Platform, see Chromeleon CDS software, BioPharma Finder software, and the Ardia Platform.