The MAM 2.0 workflow requires that you have installed and configured one or both of the following MS instruments:
  • Orbitrap Exploris 240 mass spectrometer
  • This MS instrument has full-scan MS and MS2 capability and can be applied for both, characterization and attribute monitoring.
  • Orbitrap Exploris MX mass detector
  • This MS instrument has full-scan capability and can be applied for attribute monitoring.
Follow the operating manual and the Tune Software Manual to complete installation and configuration of your MS instruments.