After you completed the research and development phase of the MAM 2.0 workflow, you are ready to transfer the PQA monitoring method to routine quality and control. With the Chromeleon CDS software, you can automate the creation of new injection sequences, so that there is no need to configure each PQA monitoring experiment individually.
Chromeleon CDS software provides automated creation of new injection sequences by the means of eWorkflow procedures. These can create new sequences that have the required structure, the correct auto-reporting and electronic signature settings, and that they contain the right instrument and processing methods, report templates, view settings, and so on.