Suggested recovery actions
- For some issues, restarting the application is the appropriate recovery action.
- As a fix, we generally do not recommend reinstalling the software or the operating system, which more commonly occurs after you install a new hard drive.
- After the installation on the software on some PCs, there is an issue with viewing the IQ report using the displayed link. If this occurs, the IQ report can be found in
C:\Program Files\Thermo\IQ Reports.
Feature requests and other omitted items
- We do not include issues where there is insufficient information logged to successfully reproduce the reported problem.
- We do not list feature requests as software issues, regardless of the reported significance or severity of the request. Product managers evaluate logged feature requests for future releases.
- We report only discrepancies in the documented software as known issues.
Terminology
The following tables define the terms used to describe the severity and risks associated with the known defects detailed in the Known Defects table. Product management assesses risk, which can differ significantly from the reported severity.
Severity | Interpretation |
|---|---|
Critical | A problem that renders the system unusable because either an entire function is unusable and no workaround exists or use of the current system compromises data integrity or results in data loss. Catastrophic problems also include significant and non-obvious quantitative errors and all human and instrument safety issues. |
High | A serious issue that does not affect data integrity (meaning data loss, corruption of data, or the wrong answer), but affects the customer’s ability to use the product as designed. It can be a failure, design issue, or documentation error or omission. A workaround might or might not exist. |
Medium | A minor error or poor behavior of a product feature. There is likely a workaround. |
Low | An issue that has a limited effect on customer usage of the product, including defects with visibility so low that a customer might never see it or ease-of use-issues or other items not causing any performance degradation. |
Risk | Interpretation |
|---|---|
High | Occurrence is likely to happen and can compromise operation. |
Medium | Occurrence is uncommon, but if it occurs, can compromise operation. |
Low | Issue is minor; however, the software might operate differently from a user’s expectations. A workaround is often available. |
No risk | This issue causes no problems but is a common inconsistency or an issue with a low severity level. |
Known defects
The following table lists the known defects in the Isotope Discoverer 1.0 application, categorized by software section, including a brief description and information related to each defect’s severity and risk. The item ID is the internal tracking number assigned to each issue.
Severity | Risk | Item ID | Software area | Description |
|---|---|---|---|---|
Medium | Medium | 943570 | Results tables | The "Is extraction incomplete" parameter displays False despite being True. |
Low | Low | 927824 | Lists & Libraries view | Users cannot import isotopes from |
Low | No risk | 943568 | Instrument compatibility Data analysis | Isotopocule extraction fails for raw files acquired on Orbitrap Tribrid™ and Astral™ series instruments. |
Low | No risk | 943573 | User interface | For some display settings, workflow nodes are not displayed at the cursor when dragged into the workflow tree. |