End-to-end isotope analysis workflow
The Isotope Discoverer release introduces a complete processing workflow for mass spectral isotope analysis, guiding users from raw-data isotopocule extraction through filtering, annotation, summarization, drift correction, calibration, and big-delta calculation.
Flexible targeted and exploratory data extraction
Users can extract known compounds and isotopocules using defined isotopocule tables or perform untargeted extraction to explore the most intense peaks without predefined targets.
Guided workflow creation, validation, and job management
A visual workflow editor, reusable workflow templates, guided study creation, setup validation, and job queue controls help users configure, submit, monitor, prioritize, and manage analyses more efficiently.
Integrated result review, visualization, and raw-data inspection
The application provides dedicated result tables, raw-file chromatogram and mass-spectrum views, and multiple charting options—including scatter, histogram, bar, pie, ẟ versus ẟ, and statistics box-and-whisker charts—to support interactive data review.
Export, reporting, statistics, and expert extensibility
Users can export result tables, charts, and workflow outputs; generate report documents from templates; run Allan analysis and descriptive statistics; fine-tune advanced evaluation parameters; and extend processing with custom post-processing scripts.