Isotope Discoverer is a study-based software application for high-precision isotopologue and isotope ratio analysis of high-resolution accurate-mass (HRAM) data acquired with Thermo Scientific Isora™ mass spectrometers. It provides specialized workflows for evaluating isotope ratios across multiple elements, isotopologues, and compounds using specialized acquisition modes and advanced sample-introduction techniques.
The application organizes raw data, processing results, and isotope ratio outputs within a structured study environment, supporting traceable and reproducible data analysis. Dedicated visualization and evaluation tools facilitate method development, data quality assessment, and interpretation of isotope ratio results.
TIP
For more information, refer to the Isotope Discoverer User Guide embedded in the application or available on the Thermo Fisher Scientific Chromatography and Mass Spectrometry Help Portal.
NOTE
In these Release Notes, the terms isotopocules and isotopologues are treated as synonyms.
For consistency, the Isotope Discoverer application uses the term isotopocules throughout.
In the scientific literature, these terms are not always used interchangeably and may have different meanings depending on the discipline and context.
These release notes describe the features and known limitations for the initial release of Isotope Discoverer.