The Isotope Discoverer 1.0 application can process data files produced by high-resolution accurate-mass (HRAM) Thermo Scientific™ instruments, such as the Orbitrap Isora™ , Orbitrap Exploris™, Orbitrap Excedion™, Q Exactive™, and Exactive™ series of mass spectrometers. However, full support is only available for Orbitrap Isora, which offers a fully integrated solution for isotope ratio analysis and data processing.
At present, Isotope Discoverer is only available with Orbitrap Isora and not sellable as a standalone software product.
The Isotope Discoverer 1.0 application is not compatible with data files acquired on Orbitrap Astral™ and Orbitrap Tribrid series (Such as Orbitrap IQ-X™, Orbitrap ID-X™, Orbitrap Fusion™) mass spectrometers.
The following table shows a detailed instrument compatibility matrix of the Isotope Discoverer 1.0 application.
Instrument platform | Basic isotope ratio and δ calculation | Full feature support | Comments |
|---|---|---|---|
Orbitrap Isora™ series | Supported | Supported | Fully supported |
Orbitrap Excedion™, Orbitrap Exploris™, and Q Exactive™/Exactive™ series | Compatible | Compatible | Compatible |
Orbitrap Exploris™ GC | Compatible | Not supported | The Average by Valve Position node might not operate as intended. |
Chromeleon™ chromatography data system | Compatible | Not supported | Valve position export and raw file metadata are not available. |
Orbitrap Astral™ series | Not supported | Not supported | Isotopocule extraction is not supported because of the different raw file format. |
Orbitrap Tribrid™ series | Not supported | Not supported | Isotopocule extraction is not supported because of the different raw file format. |
The following table lists the minimum and recommended hardware and software configurations required for Isotope Discoverer 1.0 operation.
System | Minimum | Recommended |
|---|---|---|
Hardware |
|
|
Software |
| |