NOTE
The Export Filter subpage of the processing method editing wizard is only available when running BioPharma Finder in local mode.
The automatic intact report workbook generation feature is not available whenconnected to the Ardia Platform.
The Export Filter subpage of the processing method editing wizard displays the sequence-specific and component-based filtering parameters you can apply to an intact report workbook.
NOTE
The Export Filter subpage of the processing method editing wizard is active even if you do not select the Enable Export for External Reporting checkbox on the Intact Mass Analysis page.
However, to automatically create an intact report workbook, you must select the Enable Export for External Reporting checkbox.
Figure Export Filter subpage navigation
1234
No.DescriptionNo.Description
1
Parameters tab (processing method editing wizard)
3
Sequence-Specific Filters area
2
Subpage navigation bar
4
Component-Based Filters area
The following table describes the parameters on the Export Filter page.
Table Export Filter subpage parameters
Parameter
Description
Sequence-Specific Filters (only applies if a sequence is selected during the analysis)
Include components that are:
  • Identified (checkbox): Select this checkbox to include identified components in the exported intact report workbook.
  • Default: Checked
  • Unidentified (checkbox): Select this checkbox to include unidentified components in the exported intact report workbook.
  • Default: Checked
Range for Matched Mass Error (checkbox)
Select this checkbox to only include components within a Matched Mass Error range in the exported intact report workbook.
Default: Checked
If checked, enter a numeric value for the lower and upper range limits in the boxes and select a unit (ppm or Da) from the dropdown list.
Note: The default Range for Matched Mass is set by the Sequence Matching Mass Tolerance parameter on the Identification subpage, but can be changed here without affecting the Sequence Matching Mass Tolerance parameter.
  • Lower limit: −20.00 (Range = 0.00 to −999,999,999.99)
  • Upper limit: 20.00 (Range = 0.00 to 999,999,999.99)
  • Unit: ppm (Options: ppm or Da)
Component-Based Filters
Relative Abundance Threshold (%)
Select this checkbox to only include components over a Relative Abundance Threshold (%) in the exported intact report workbook.
Default: Checked
If checked, enter a numeric value for the Relative Abundance Threshold (%) in the box.
Default: 1.0%
Range: 0.0 to 100.0%
Top number of components
Enter the maximum number of components to include in the exported intact report workbook.
Default: 50
Range: 0 to 50
Note: If the export filters for the intact report workbook result in more than 50 components, only the top 50 components (ranked by relative abundance) are included.