The tables below list the sources for the report elements (tables, plots, and sequence coverage map) available in the BioPharma Finder Report utility.
Report element | Source of information | Workflow |
---|---|---|
Consolidated Results table | Process and Review page > Consolidated Results table |
|
Charge State Results table | Process and Review page > Charge State Results table |
|
Results table | Process and Review page > Results table (component (top) level) |
|
HCP Quantitation table | Mapping > Host Cell Protein Quantitation page > Host Cell Protein Quantitation results table |
|
Main Modification Summary table | Mapping > Modification Summary page > Modification Results (lower table) |
|
Supplementary Results table | Mapping > Modification Summary page > Modification Results (upper table) |
|
Data Analysis Summary table | Load Results page > Information from the table of experiments |
|
Experimental Data table | Parameters > Save Method page > Experimental Data table |
|
Protein Sequence, Protease & Modification table | Parameters > Component Detection page parameters |
|
Sequence, mRNA & Modification table | Parameters > Component Detection page parameters |
|
Sequences & Modification | Parameters > Component Detection page parameters |
|
Identification table | Parameters > Identification page parameters |
|
Component Detection table | Parameters > Component Detection page parameters |
|
Host Cell Protein table | Parameters > Host Cell Protein page parameters |
|
Hydrogen Deuterium Parameters table | Parameters > Hydrogen Deuterium Exchange page parameters |
|
Matched Charges table | Process and Review page > Results table (charge state (lowest) level) |
|
Average DAR table | Process and Review page > Average DAR table |
|
UV Analog trace | Process and Review page > Chromatogram pane |
|
Total Ion Chromatogram (TIC) | Process and Review page > Chromatogram pane |
|
Base Peak Chromatogram (BPC) | Process and Review page > Chromatogram pane |
|
Extracted Ion Chromatogram (XIC) | Process and Review page > Chromatogram pane |
|
Mixed Traces (single raw file only) | Process and Review page > Chromatogram pane |
|
Abundance Trace (Sliding Windows only) | Process and Review page > Chromatogram pane |
|
Deconvoluted Spectrum | Process and Review page > Deconvoluted Spectrum pane |
|
Deconvoluted Spectrum (Component) | Process and Review page > Deconvoluted Spectrum pane |
|
Source Spectrum | Process and Review page > Source Spectrum pane |
|
Base Peak Chromatogram Peak Shading | Mapping > Coverage page > Chromatogram pane |
|
Protection Factor plot | Mapping > Hydrogen Deuterium Exchange page > Protection Factor plot |
|
Sequence Coverage Map | Mapping > Coverage page > Sequence Coverage Map pane |
|