Prerequisites
- You have loaded experiment results from the Ardia Platform data repository.
- You are viewing the Design page on the Report tab.
Procedure
- On the Design page, under the Insert tab, select Sequence Coverage Map.
- The Select Raw File window opens.
- In the Select Raw File window, select the raw data file you want to source for the sequence coverage map from the dropdown list.
- Below, select the Coverage Map Type: Default or User Defined and then select OK.
- The sequence coverage map is inserted in the report.
TIP
For more information on displaying user-defined sequence components on the Sequence Coverage Map, refer to Select Sequence Coverage Map components.
IMPORTANT
If a "No data available..." message is displayed on the report sheet, manually source the sequence coverage map.
- If necessary, source the information for the sequence coverage map as follows:
- In the BioPharma Finder navigation bar, select the Mapping tab.
- On the Mapping page, in the Sequence Coverage Map pane, select the Add to Report button at the top left of the pane.
- The application displays the Adding to Report Progress bar while the information is sent to the report.
- When the results have been loaded to the report, select the Report tab and then select the Refresh Data button at the top right of the page.
- The application updates the sequence coverage map with the sourced experiment results.