Prerequisites
- You have loaded experiment results from the Ardia Platform data repository.
- You are viewing the Design page on the Report tab.
Procedure
- On the Design page, under the Insert tab, select Report Table.
- The Insert Report Table window opens, displaying all the tables available to add to the report.
Figure Insert > Report Table (for Peptide Mapping Analysis) options - In the Insert Report Table menu, select the table you want to insert.
- The selected table (blank) is added to the current report sheet.
IMPORTANT
Before you manually source the information for the report elements, some elements might appear blank and/or a "No data available..." message might be displayed on the report sheet.
- Next, source the appropriate information for the table as follows:
- In the BioPharma Finder navigation bar, navigate to the page that contains the table you added to the report.
- In the table, select the checkbox for the component results you want to include in the associated report table.
- Right-click anywhere in the table and select Add to Report from the shortcut menu, and then select All Results or Checked Results from the submenu as appropriate.
- The application displays the Adding to Report Progress bar while the information is sent to the report.
- When the results have been loaded to the report, select the Report tab and then select the Refresh Data button at the top right of the page.
- The application updates the report table with the sourced experiment results.
IMPORTANT
When you select components to include as results in the report, the following behavior occurs:
For Peptide Mapping Analysis and Oligonucleotide Analysis reports, the selected components are used as source information for only results tables added to the report. Other report elements must be sourced manually.
Furthermore, for Peptide Mapping Analysis reports, the Consolidated Results table and Charge State Results table must be sourced individually. For Oligonucleotide Analysis reports, the Consolidated Results table and Charge State Results table are simultaneously sourced when components from either table are added to the report.
For Intact Mass Analysis reports, the selected components are used as source information for all report elements automatically.
TIP
When you add a report table, the Report Table Tools subtab appears. Use this tab to customize the table appearance and sort and filter data as needed.
Figure Report Table Tools subtab TIP
Depending on the report table, some available data columns might not be initially included in the table. You can manually insert additional data columns needed.
- To insert additional data columns, do the following:
- Right-click anywhere in the body of the report table and select Insert Column from the shortcut menu.
- From the Insert Report Table Column dropdown menu, select the data column you want to add.
- The application adds the selected data column to the report table.
NOTE
If you add a data column that does not apply to the current experiment, the column will appear blank in the report table.