After you add a sequence and specify processing parameters, generate theoretical components by selecting the Process button in the Theoretical Protein/Peptide Manager. The Results table displays the theoretical component results and you can save all or selected results as a target peptide workbook or other file type as needed.
Generate theoretical component results
Prerequisites
- You are viewing the Theoretical Protein/Peptide Manager.
- You added a protein sequence to the Theoretical Protein/Peptide Manager.
- You specified digestion and target m/z processing parameters and added modifications to the sequence, as necessary.
Procedure
- In the Theoretical Protein/Peptide Manager, select the Process button to generate theoretical components.
- On the right side of the page, the Results table displays the theoretical component results.
- You can reprocess all or selected results as needed, depending on the processing mode selected on the Add/Edit Target m/z tab.
Save theoretical components to a target peptide workbook
TIP
For more information on creating and managing BioPharma Finder workbooks, see Introduction to BioPharma Finder workbooks.
Prerequisites
- You have generated theoretical components using the Theoretical Protein/Peptide tool.
Procedure
- (Optional) In the Theoretical Protein/Peptide Manager Results table, select the checkbox in the row of each component that you want to save.
IMPORTANT
You can save a maximum of 250 components to a target peptide or oligo workbook.
- To save results to a target peptide workbook, select Save As Peptide Workbook on the command bar.
- Select one of the following submenu commands:
- All to export all components in the table to a workbook.
- Checked to export only the selected components to a workbook.
- The Save Peptide Workbook As dialog box opens.
- In the Save Peptide Workbook As dialog box, select one of the following options:
- Create a New Peptide Workbook to add the components to a new workbook.
- Select an Existing Workbook to add the components to an existing workbook.
- (For the Create a New Peptide Workbook option) In the Workbook Name box, type a name for the workbook.
TIP
Workbook names can contain alphanumeric, space, underscore (“_”), and period (“.”) characters.
IMPORTANT
You can overwrite an existing workbook by entering the same workbook name.
- (For the Select an Existing Workbook option) In the table, select the checkbox for an existing workbook you want to update.
- (Optional) In the Description box, type a description for the workbook.
- In the Number of Isotopes per Peptide box, specify the number of isotopes you want to include for the selected components.
- Select Save.
- The application saves the selected components to a new or existing target peptide workbook, as indicated.
Save theoretical components as a different file type
TIP
Save your theoretical component results as an .msqc file to open the results in the BioPharma Finder application later.
Prerequisites
- You have generated theoretical components using the Theoretical Protein/Peptide tool.
Procedure
- (Optional) In the Results table, select the checkbox in the row of each component that you want to save.
- To save results to an Excel (.xlsx) or BioPharma Finder (.bpf) file, select Export Results on the command bar.
- Select one of the following submenu commands:
- All to export all components in the table.
- Checked to export only the selected components.
- Then, select one of the following additional submenu commands:
- As Displayed to export the components as an Excel file (.xlsx).
- Chromeleon to export the components as a BioPharma Finder file (.bpf).
- To save all components as a data file (.msqc), select Save Results As on the command bar.
- The Save As dialog box opens.
- In the Save As dialog box, browse to the folder location where you want to save the file, type a name for the file, and select Save.
- The file containing the theoretical component results is saved in the specified file location.