The following table describes the information in the Results table on the Workbook Editor page for an intact workbook.
NOTE
The information included in the Results table in an intact workbook varies depending on the selected deconvolution algorithm (ReSpect or Xtract), deconvolution method (average over selected retention time, automatic peak detection, or sliding windows) and other processing method parameters.
For more information, refer to View the Results table.
Column | Description |
---|---|
Component level | |
+/− | Select to show or hide the lower level of raw file information related to the current component row. |
Row number (bolded) | The number assigned to each visible workbook row in the table. This sequential numbering does not change when you sort or filter the table. |
(Checkbox) | Select this checkbox to export a workbook entry. To select or clear all of the checkboxes at once, select or clear the checkbox in the column header. If you filter the table, the following occurs:
|
Level | Indicates that the row is displaying component information (top level). |
(Various information columns) | |
Raw file level | |
+/− | Select to show or hide the lower level of raw file information related to the current component row. |
Row number (bolded) | The number assigned to each visible workbook row in the table. This sequential numbering does not change when you sort or filter the table. |
Level | Indicates that the row is displaying raw file information (middle level). |
Raw File Name | Displays the name of the raw data file. |
Condition | Displays the condition assigned to the raw data file (if any). |
(Various information columns) | |
Charge state level | |
Row number (bolded) | The number assigned to each visible charge state row in the table. This sequential numbering does not change when you sort or filter the table. |
Level | Indicates that the row is displaying charge state information (lower level). |
Charge State | Displays the charge state for the component. |
(Various information columns) |