The following table describes the information in the Workbook Manager table.
Table Workbook Manager table information
Column
Description
Row number (bolded)
The number assigned to each workbook row in the table. This sequential numbering does not change when you sort or filter the table.
(Checkbox)
Select the checkbox for a workbook row to select it.
Name
Displays the name of the workbook.
Category
(Target peptide workbooks only) Displays the category of the protein sequence used to generate the results saved to the workbook.
Creation Date and Time
Displays the date and time when you created the workbook.
Last Modified Time
Displays the date and time when you last modified the workbook.
Number of Entities
Displays the number component masses saved to the workbook.
Number of Groups
(Target peptide workbooks only) Displays the number of Relative Quantitation Groups in the workbook.
Deconvolution Algorithm
(Intact workbooks only) Displays the deconvolution algorithm used to generate the results saved to the workbook.
Source Spectrum Method
(Intact workbooks only) Displays the method used to generate the source spectrum for the results saved to the workbook.
Associated Experiment
(Intact workbooks only) Displays the name of the experiment that generated the results saved to the workbook.
Associated Processing Method
(Intact workbooks only) Displays the name of the processing method used to generate the results saved to the workbook.
Associated Sequences
(Intact workbooks only) Displays the name of the sequences used to generate the results saved to the workbook.
Description
Displays the description you entered (if applicable) when you saved the workbook.