The following table describes the commands in the Workbook Manager.
Command | Description |
---|---|
New (Oligo Calculator and Workbook page only) | Opens a new oligo workbook where you can generate and save theoretical sequences. |
Edit | Opens the appropriate workbook editor so you can edit workbook parameters and components. |
Delete | Deletes the selected workbooks in the Workbook Manager table. |
Import | Opens a dialog box where you can browse and select |
Export | Exports the selected workbooks in the Workbook Manager table as |