An oligo workbook contains saved data that can be shared with the Chromeleon chromatography data system. You can save selected experiment results or generate and save theoretical components to an oligo workbook.
IMPORTANT
You can save a maximum of 250 components to a target peptide or oligo workbook.
In the Oligonucleotide Calculator and Workbook Editor, you can change the charge state, m/z, and mass ranges, as well as the number of isotopes displayed and update the results as needed.