A workbook is a saved set of data containing the results from an experiment. Workbook files (.wbpf) can be shared with the Chromeleon chromatography data system (CDS) for further analysis.
The following table describes the types of workbooks you can create in BioPharma Finder.
BioPharma Finder Workbooks
Workflow
Workbook
Use case
Peptide Mapping Analysis
target peptide workbook
  • Run targeted Peptide Mapping Analysis experiments in BioPharma Finder
  • Export selected results/theoretical components to the Chromeleon CDS or as an Excel file
Oligonucleotide Analysis
oligo workbook
  • Export selected results/theoretical components to an Excel file
Intact Mass Analysis
intact workbook
  • Export selected components and associated processing parameters to the Chromeleon CDS or as an Excel file