Procedure
- On the BioPharma Finder Home page, in the Experiment Types pane, select the appropriate workflow link.
- The corresponding workflow page opens.
- In the [Workflow] Definition area, type the name of the experiment in the Experiment Name box.
TIP
Experiment names can contain up to 50 characters, including alphanumeric, space, underscore (“_”), and period (“.”) characters.
IMPORTANT
You can overwrite a completed experiment by entering the same experiment name.
You cannot overwrite an experiment that is currently submitted to the run queue or is actively processing or canceling in the run queue.