You can run the BioPharma Finder application in local mode or connected to the Ardia Platform. When you launch the application for the first time, it opens by default in local mode, disconnected from the Ardia Platform.
In both local and Ardia Platform mode, the BioPharma Finder Home page displays the Experiment Types navigation pane and workflow links.
The Experiment Types navigation pane contains five tabs that open the Sequence Manager or one of the following workflows:
- Peptide Mapping Analysis
- Oligonucleotide Analysis
- Intact Mass Analysis
- Top Down Analysis