The following table describes the commands in the Oligonucleotide Calculator and Workbook Editor.
Command | Description |
|---|---|
Command bar | |
Import Sequence | Imports multiple oligonucleotide sequences simultaneously from a specially-formatted |
Export | Exports the currently open oligo workbook as an Excel ( |
Save | Saves the oligo workbook to the Workbook Manager. |
Close | Closes the Oligonucleotide Calculator and Workbook Editor. |
Results table | |
Delete All | Deletes all components in the Results table. |
Delete Checked | Deletes the selected components in the Results table |
Simulated and Predicted MS2 Spectra panes | |
Reset Scale | Restores the original scale to the pane. |
Copy | Copies the image viewable in the pane, including labeling and shading. |