Prerequisites
  1. You have processed an experiment.
  2. You are viewing the Process and Review page.
Procedure
  1. On the Process and Review page, select the checkbox in the row of each component in the Charge State Results table that you want to save to a workbook.
  2. TIP
    To select all rows in the table, select the checkbox in the column header.
  3. NOTE
    You cannot save unidentified components or components with "0" charge state, unspecified modifications, dimers, adducts, or any type of gas phase modifications to the workbook
    Clear the checkboxes for these components to save the workbook.
  4. IMPORTANT
    You can save a maximum of 250 components to a target peptide or oligo workbook.
  5. Right-click anywhere in the Charge State Results table and select Save As Oligo Workbook from the shortcut menu.
  6. Select one of the following submenu commands:
  7. All to export all results in the Charge State Results table to a workbook.
  8. Checked to export only the selected results to a workbook.
  9. The Save Oligo Workbook As dialog box opens.
  10. In the Save Oligo Workbook As dialog box, select one of the following options:
  11. Create a New Oligo Workbook to add the components to a new workbook.
  12. Select an Existing Workbook to add the components to an existing workbook.
  13. (For the Create a New Oligo Workbook option) In the Workbook Name box, type a name for the workbook.
  14. TIP
    Workbook names can contain alphanumeric, space, underscore (“_”), and period (“.”) characters.
  15. IMPORTANT
    You can overwrite an existing workbook by entering the same workbook name.
  16. (For the Select an Existing Workbook option) In the table, select the checkbox for an existing workbook you want to update.
  17. (Optional) In the Description box, type a description for the workbook.
  18. In the Number of Isotopes per Oligo box, specify the number of isotopes you want to include for the selected components.
  19. Select Save.
  20. The application saves the selected components to a new or existing oligo workbook, as indicated.