Prerequisites
- You have registered with the Ardia Platform.
- You have completed a Peptide Mapping Analysis, Oligonucleotide Analysis, or Intact Mass Analysis experiment.
- You have saved the experiment to the Ardia Platform data repository.
Procedure
- Start the BioPharma Finder application in Ardia Platform mode.
- On the BioPharma Finder Home page, in the Experiment Types pane, select the appropriate workflow link.
- On the navigation bar, select the Load Results tab.
- The Load Results page opens, displaying the Ardia Platform Data Explorer.
Figure Load Results page (connected to Ardia Platform) No. Description No. Description 1
Load Results tab
3
Ardia Platform data repository folder tree
2
Command bar
4
Ardia Platform data repository table
- In the left pane of the Load Results page, navigate to your experiment folder in the Ardia Platform data repository and then select the folder.
- In the right pane, select the checkbox for the experiment that you want to open.
- Select Load Results on the command bar of the Load Results page.
- The experiment opens, displaying the Process and Review page.
- On the Process and Review page, in the Results table, select the checkbox for the components you want to include in the report.
- Right-click anywhere in the Results table and select Add to Report from the shortcut menu, and then select All Results or Checked Results from the submenu as appropriate.
IMPORTANT
When you select components to include as results in the report, the following behavior occurs:
For Peptide Mapping Analysis and Oligonucleotide Analysis reports, the selected components are used as source information for only results tables added to the report. Other report elements must be sourced manually.
Furthermore, for Peptide Mapping Analysis reports, the Consolidated Results table and Charge State Results table must be sourced individually. For Oligonucleotide Analysis reports, the Consolidated Results table and Charge State Results table are simultaneously sourced when components from either table are added to the report.
For Intact Mass Analysis reports, the selected components are used as source information for all report elements automatically.
- Select the Report tab on the navigation bar.
- The Design page opens, showing a cell grid arrangement similar to an Excel spreadsheet.
TIP
The BioPharma Finder Report utility has the same cell grid arrangement and similar features and commands found in an Excel spreadsheet.
Figure Report page, showing the Design subpage (blank) - To create the report from a template, do the following:
- On the Design page, under the File tab, select New.
- In the New Report Template window, select a report template from the options displayed.
- The Design page displays the report template containing the appropriate default elements.
NOTE
Report templates might contain multiple sheets. To navigate the report pages, select a sheet tab at the bottom left of the Design page.
- Alternatively, you can manually add report elements one at a time, as follows:
- Insert a new sheet
- Insert a report table
- Insert a report plot
- Insert a report variable
- Insert a sequence coverage map
IMPORTANT
Before you manually source the information for the report elements, some elements might appear blank and/or a "No data available..." message might be displayed on the report sheet.