The Sequence Manager collates all protein and oligonucleotides sequences added to the application.
TIP
You can access the Sequence Manager by selecting the Sequence Manager link on the BioPharma Finder Home page.
Use the Sequence Manager to import, export, and delete protein and oligonucleotide sequences in the BioPharma Finder application. In the Sequence Manager, you can access the Host Cell Protein Database Manager and Theoretical Protein/Peptide Manager subpages as needed.
Figure Sequence Manager navigation
123456789
No.DescriptionNo.Description
1
Open a file browser to select and import BioPharma Finder sequence files (.sequencebpf)
6
Delete the selected sequences
2
Export the selected sequences as BioPharma Finder sequence files (.sequencebpf)
7
Open the Host Cell Protein Database Manager to add or delete HCP databases the application uses to detect and quantify trace contaminants
3
Open the Protein Sequence Editor to add and customize a new protein
8
Open the Theoretical Protein/Peptide Manager to generate and save theoretical components for a protein sequence
4
Open the Oligonucleotide Sequence Editor to add and customize a new oligonucleotide
9
The Sequence Manager table displays all protein and oligonucleotide sequences added to the application
5
Open the appropriate Sequence Editor to edit the selected sequence