The Save Method page contains two sections: Method Name/Description (editable) and a read-only section summarizing experimental data, sequences and modifications, and processing parameters.
| No. | Description | No. | Description |
|---|---|---|---|
1 | Parameters tab (processing method editing wizard) | 4 | Experiment information report area |
2 | Subpage navigation bar | 5 | Finish button |
3 | Method Name and Description area |
Once you have entered a method name, select the Finish button to save the method and return to the Intact Mass Analysis submit the experiment for processing.
IMPORTANT
You cannot overwrite a default processing method. You must use a new name to save changes made to a default method.