In the Oligonucleotide Sequence Editor, you can change the 5'-terminal, 3'-terminal, base, backbone linker, and 2'-ribose groups for an entire oligonucleotide sequence or within a specific chain using the Edit Sequence pane.
TIP
You can create custom oligo building blocks and assign them to a sequence using the Edit Sequence pane.
Prerequisites
- You are viewing the Oligonucleotide Sequence Editor.
Procedure
- In the Oligonucleotide Sequence Editor, expand the Edit Sequence pane on the right side of the page.
- In the Select Chain dropdown list, do either of the following:
- To apply global changes to all oligo building blocks, select All Chains.
- To apply changes to a specific chain only, select the appropriate chain number.
Figure Edit Sequence pane showing Select Chain options - If you selected All Chains, do the following:
- Select a subunit from the Subunit dropdown list.
- In the From dropdown list, select the building block you want to change.
- In the To dropdown list, select the new building block you want to use for the selected subunit.
- Select Apply.
- The Oligo Sequence Map updates and displays the entire oligonucleotide sequence comprised of the specified building blocks.
- The Sequence Information pane updates and displays the recalculated Target Oligonucleotide and Chain parameters.
- If you selected a specific chain, do the following:
- Above the table, in the 5'-Terminal dropdown list, select the 5'-terminal for the specified chain.
- In the table, use the dropdown lists to specify the backbone linker, base, and 2'-ribose building blocks for each triplet in the specified chain.
- Below the table, in the 3'-Terminal dropdown list, select the 3'-terminal for the specified chain.
- Select Apply.
- The Oligo Sequence Map updates and displays the chain comprised of the specified building blocks.
- The Sequence Information pane updates and displays the recalculated Target Oligonucleotide and Chain parameters.
- Save the oligonucleotide sequence.