Procedure
- On the BioPharma Finder Home page, in the Experiment Types pane, select the Sequence Manager link.
- The Sequence Manager opens.
- On the right side of the Sequence Manager, select the New Oligonucleotide button.
- The Oligonucleotide Sequence Editor opens.
- In the Oligonucleotide Sequence Editor, select Import Fasta File on the command bar.
- The Open dialog box opens.
- Locate and select the FASTA file (.fasta) you want to import and select Open.
NOTE
FASTA files must not exceed 1 MB for import.
- If the entered chain contains invalid oligo building blocks or an invalid format, an error message appears.
- The Oligo Sequence Map on the left side of the page updates and displays the added chain.
- The name of the chain appears above the chain sequence, indicated by a greater-than sign (>) to distinguish each chain.
- Additionally, the read-only information fields (Monoisotopic Mass, Average Mass, and Formula) in the Sequence Information pane > Target Oligonucleotide area update to reflect the properties of the entire protein.
- In the Sequence Information pane > Chain area, you can view the Monoisotopic Mass and Average Mass properties for an individual chain by selecting the chain number from the dropdown menu.
- On the left side of the Oligonucleotide Sequence Editor, in the Sequence Information pane > Target Oligonucleotide area, enter the following information:
- Name: Type the name of the oligonucleotide sequence.
- Description: (Optional) Type a description for the oligonucleotide sequence.
- Category: Select the category for the oligonucleotide sequence from the dropdown menu.
- Save the oligonucleotide sequence.
- The application adds the saved sequence to the Sequence Manager table.