The Identification subpage of the processing method editing wizard contains six sections: Sequence Matching Mass Tolerance, Enable Drug-to-Antibody Ratio (Protein Only), Select a variable modification candidate for the DAR calculation, Multiconsensus Component Merge, Sequences Added to Experiment (table), and Global Sequence Reference (table).
As needed, you can add, delete, or edit sequences for the experiment using the Sequences Added to Experiment and Global Sequence Reference tables.
Figure Identification subpage navigation
12345678
No.DescriptionNo.Description
1
Parameters tab (processing method editing wizard)
5
Select a variable modification candidate for the DAR calculation area
2
Subpage navigation bar
6
Multiconsensus Component Merge area
3
Sequence Matching Mass Tolerance parameter
7
Sequences Added to Experiment table
4
Enable Drug-to-Antibody Ratio (Protein Only) checkbox
8
Global Sequence Reference table
The table below describes the parameters on the Identification subpage.
Table Identification subpage parameters
Parameter
Description
Sequence Matching Mass Tolerance
Specifies the mass tolerance (in ppm or Da) within which the mass of the full sequence and individual components must fall to be considered a match.
Enable Drug-to-Antibody Ratio (checkbox)
(Protein only) Prompts the application to determine the average drug-to antibody ratio (DAR) value based on the drug load assignments to the identifications. You must select a modification candidate.
Select a variable modification candidate for the DAR calculation
(Protein only) These parameters show the information for the selected variable modification candidate.
List of Modifications (drop-down list)
(Enabled only when you select the Enable Drug-to-Antibody Ratio checkbox) Lists all of the side chain variable modifications available in the Protein Sequence Editor.
An N-terminal or a C-terminal modification that is not already included as a side chain modification is not available in this list. If you want to select a missing modification, first add it as a custom modification to the list of side chain modifications using the Protein Sequence Editor. See Assign modifications to a protein sequence.
Select one modification from this list to be the drug linker. The residue, monoisotopic mass, and average mass information for the selected modification appears.
When you select a drug linker from this list, after the application finishes the matched sequence identification step, it automatically searches for all of the modifications that match the selected drug linker and then reports the number of matched modifications as the Drug Load value on the deconvoluted spectra and in the Matched Sequence pane of the Process and Review page.
If you select the Enable Drug-to-Antibody Ratio checkbox but do not select a modification from this list, the following occurs:
  • If you manually process the experiment, the application displays an error message informing you that a modification selection is required.
  • If you automatically process the experiment, the application does not display an error message, but the resulting drug load values appear as 0.
Residue
(Not editable) Residue of the selected modification.
Monoisotopic Mass
(Not editable) Monoisotopic mass of the selected modification.
Average Mass
(Not editable) Average mass of the selected modification.
Multiconsensus Component Merge
These parameters control the merging of the deconvoluted results for multiple raw data files when you select Multiconsensus as the result format on the Intact Mass Analysis page.
Deconvolution Mass Tolerance
Specifies the mass tolerance (in ppm or Da) that components from individual raw data files must meet to be merged into a multiconsensus component.
The application excludes individual raw data file components whose masses differ by more than this tolerance from a multiconsensus component.
RT Tolerance
Specifies the maximum difference in retention time (RT) that components from individual files must meet to be merged into a multiconsensus component.
The application excludes individual raw data file components whose retention times are separated by more than this gap from a multiconsensus component.
Minimum Number of Required Occurrences
Specifies the minimum number of individual raw data files that must be included in a multiconsensus component for it to be considered valid.
The application excludes multiconsensus components comprised of fewer than this number of files.