On the Load Results page of any workflow, the software version used to create the experiment is displayed in the Software Version column of the Load Results table.
If you are using BioPharma Finder 5.0, you cannot open experiments that were created in updated versions of the software from the Ardia Platform data repository.
If you open and edit an experiment created in BioPharma Finder 5.0 using an updated version of the software, you must save the experiment as a new experiment. You cannot overwrite the older existing file.
When you save an experiment created in a previous version as a new experiment in BioPharma Finder 5.5, the application automatically adds following comment in the Save As dialog box: "This result was originally generated from BPF [version]".