Insert a report variable - BioPharma Finder Software - Intact Mass Analysis User Guide - Insert a report variable - Report: Design page navigation - Reporting - BioPharma Finder Software - BioPharma Finder Software
Intact Mass Analysis User Guide
Product Tree
Ardia Platform > Software > BioPharma Finder Software
Desktop Software > BioPharma Finder Software
Document Type
User Guide
Software version
5.3
You have loaded experiment results from the Ardia Platform data repository. You are viewing the Design page on the Report tab. On the Design page, select a cell in the cell grid on the report sheet where you want to insert a report variable. Under the Insert tab, select Report Variable and then select General Information . The Insert Report Variable window opens, displaying a dropdown list of report variables. Figure Insert Report Variable windowIn the Insert Report Variable window, select a report variable from the dropdown list, and then select OK . The selected report variable is inserted in the report.