Prerequisites
  1. You have loaded experiment results from the Ardia Platform data repository.
  2. You are viewing the Design page on the Report tab.
Procedure
  1. On the Design page, do either of the following:
  2. Under the Insert tab, select New Sheet.
  3. At the bottom of the Report page, right-click on the current sheet tab and select Insert from the shortcut menu
  4. At the bottom of the Report page, Select the New Sheet button (+).
  5. The new sheet is added to the report.
  6. Figure Insert a new sheet
    123
    No.DescriptionNo.Description
    1
    New Sheet button
    3
    Add New Sheet (+) button (not visible here)
    2
    Insert Sheet option on sheet tab shortcut menu
  7. To rename a sheet, right-click on the sheet tab and select Rename from the shortcut menu. Type the sheet name and then press ENTER or click outside the tab to apply the name.
  8. To delete a sheet, right-click on the sheet tab and select Delete from the shortcut menu.
  9. To move or copy a sheet, right-click on the sheet tab and select Move or Copy from the shortcut menu.