The Deconvoluted Spectrum section of a report, shown in the following figure, displays the same spectrum that appears in the Deconvoluted Spectrum pane of the Process and Review page. See View the deconvoluted spectra.
The following table lists the parameters in the Deconvoluted Spectrum section.
Parameter | Description |
|---|---|
Relative Intensity (y axis) | Displays the ratio of the intensity of a specific peak to the intensity of the peak with the highest intensity. |
Mass (x axis) | Displays the mass of the ions formed from molecules. |