The Chromatogram section of a report, shown in the following figure, displays the chromatogram contained in the raw data file. It is the same chromatogram that appears on the Chromatogram pane of the Parameters > Component Detection or Process and Review page.
Figure Chromatogram section
Chromatogram section
The following table lists the parameters in the Chromatogram section.
Table Chromatogram section parameters
Parameter
Description
Relative Intensity (y axis)
Displays the ratio of the intensity of a specific peak to the intensity of the peak with the highest intensity.
RT (min) (x axis)
Displays the retention time (RT) of the spectrum, which is the time after injection at which a compound elutes. Retention time can also refer to the total time that the compound is retained on the chromatographic column.