The following table describes the information in the Component Specific Summary table at the bottom of the Average DAR pane on the Process and Review page.
Table Component Specific Summary table parameters
Column
Description
Row number (bolded)
The number assigned to each component row in the Component Specific Summary table.
This sequential numbering does not change when you sort or filter the table.
Drug Load
Displays the drug load values for only the components used in the DAR calculations.
Any change to the Drug Load value in the Matched Sequence pane automatically updates this column.
Protein Name
Modification
Monoisotopic Mass or Monoisotopic Mass (mean) or Average Mass or Average Mass (mean)
Matched Mass Error (ppm/Da) or Matched Mass Error (ppm/Da) (mean)
Relative Abundance
Sum Intensity or Sum Intensity (mean) or Intensity or Intensity (mean)
These columns vary depending on the number of loaded raw data files and the results format (Batch or Multiconsensus) used for the experiment.
See Results table parameters (for single file or batch format experiments) and Results table parameters (multiconsensus format experiments) for more information on these columns.