The following table describes the information in the Average Drug-to-Antibody Ratio table at the top of the Average DAR pane on the Process and Review page.
Parameter/Column | Description |
|---|---|
Experimental Average DAR | Displays the average drug-to-antibody ratio (DAR) value for the experiment, calculated as the average of all the Average DAR values from all of the raw data files loaded for the experiment. Any change to the Drug Load value automatically updates this value. |
Target Modification Name | Displays the target modification found for the matched target sequence. This is the drug linker modification that you selected in the List of Modifications in the processing method. |
Raw File Name | Displays the name or names of the raw data file or files loaded for the experiment. |
Average DAR | Displays the calculated average DAR value for each raw data file. Any change to the Drug Load value automatically updates this value here and in the deconvoluted spectra labels. |