The following table describes the information in the Target Match Sequence table at the bottom of the Matched Sequence pane on the Process and Review page.
Table Target Match Sequence table columns
Column
Description
(Check box)
Select this checkbox to view the sequence match identification in a particular table row.
You can select only one identification row in the table at a time.
When you select a different identification, the Protein/Sequence Name and Modification columns in the Intact Deconvolution Results table automatically update based on your selection.
Protein/Oligonucleotide Name
Modification
Matched Mass Error (ppm/Da) or Matched Mass Error (ppm/Da) (mean)
These columns vary depending on the number of loaded raw data files and the results format (Batch or Multiconsensus) used for the experiment.
See Additional Results table parameters for target sequence matching experiments for more information on these columns.