The following table describes the information in the Component Information table at the top of the Matched Sequence pane on the Process and Review page.
Table Component Information table rows
Row
Description
Monoisotopic Mass or Monoisotopic Mass (mean)
Average Mass or Average Mass (mean)
Sum Intensity or Sum Intensity (mean)
Intensity or Intensity (mean)
These columns vary depending on the number of loaded raw data files and the results format (Batch or Multiconsensus) used for the experiment.
See Results table parameters (for single file or batch format experiments) and Results table parameters (multiconsensus format experiments) for more information on these columns.
Drug Load
Displays the drug-to-antibody ratio (DAR) value for a specific component when the Enable Drug-to-Antibody Ratio checkbox is selected in the processing method.
The application calculates the drug load value based on the matched target sequence identification for only the identified components in the Results table.
This value is different from the Experimental Average DAR value, which is the average DAR for a complete set of measurements for an experiment, or the Average DAR value, which is the average DAR for a particular raw data file.
By default, the Drug Load value is not editable.
To change the Drug Load value for an identified component, select the Override checkbox, modify the value, and then press the ENTER key.
The application recalculates the DAR values based on your modified drug load and updates the tables in the Average DAR pane. The Results table and the drug load label on the spectra in the Deconvoluted Spectrum pane also automatically update with the modified drug load value.
Override (checkbox)
Enables the Drug Load parameter to be edited.