Right-clicking the Deconvoluted Spectrum pane on the Process and Review page opens a shortcut menu that contains the commands listed in the following table.
Table Deconvoluted Spectrum pane shortcut menu
Command
Description
Reset Scale
Restores the original scale to the pane.
Copy as Displayed
Copies the image in the pane to the clipboard as currently displayed.
Copy per Global Settings
Copies the image in the pane to the clipboard based on the global image dimensions set in the Intact Mass Analysis Settings dialog box.
Copy Data
Copies mass data (x axis) and intensity data (y axis) from the pane to the clipboard so that you can paste it into an Excel spreadsheet or another application.
For Xtract deconvolution, the copied data consists of a centroid spectrum.
For ReSpect deconvolution, the copied data consists of a profile spectrum.
Zoom Out
Shrinks the view in the pane by a factor of 2.
Zoom In
Enlarges the view in the pane by a factor of 2.
Show Identification Markers (checkbox)
Turns on and off to show or hide the markers for identified peaks.
Active only for target sequence matching experiments.
Default: On (When you close and reopen the application, the identification markers automatically appear.)
Identification Markers Color
Changes the color of the identification markers.
Active only when the Show Identification Markers command is turned on.
Select a different color from the palette, or select Advanced to enter specific RGB, HSL, or CMYK values for the new color.
Default: Orange (When you close and reopen the application, the color you previously selected reverts back to orange.)