Right-clicking the Chromatogram pane on the Process and Review page opens a shortcut menu with the commands listed in the following table.
Command | Description |
|---|---|
Reset Scale | Restores the original scale to the Chromatogram pane. |
Copy as Displayed | Copies the image in the pane to the clipboard as currently displayed. |
Copy per Global Settings | Copies the image in the pane to the clipboard based on the global image dimensions set in the Intact Mass Analysis Settings dialog box. |
Zoom Out | Shrinks the view in the pane by a factor of 2. |
Zoom In | Enlarges the view in the pane by a factor of 2. |