Right-clicking the Mirror Plot pane on the Spectra Comparison page opens a shortcut menu that contains the commands listed in the following table.
Table Mirror Plot pane shortcut menu commands
Command
Description
Reverse Order
Reverses the order of the two spectra in the Mirror Plot pane.
Reset Scale
Restores the original scale to the Mirror Plot pane.
Copy as Displayed
Copies the image in the Mirror Plot pane to the clipboard.
Copy per Global Settings
Copies the image in the Mirror Plot pane to the clipboard based on the global image dimensions set in the Intact Mass Analysis Settings dialog box.
Zoom Out
Shrinks the view in the pane by a factor of 2.
Zoom In
Enlarges the view in the pane by a factor of 2.