Create a new Intact Mass Analysis experiment on the Intact Mass Analysis page. On this page, enter the experiment name, load the raw data files, select a sequence, and select a processing method. You can run an Intact Mass Analysis experiment in Manual or Automatic mode. See Processing modes: Automatic or Manual.
IMPORTANT
For Intact Mass Analysis experiments, only protein sequences assigned to the “Intact Protein” category and oligonucleotide sequences assigned to the “Intact Deconvolution” category appear in the table.
If your sequence does not appear in the table, you can change its assigned category to “Intact Protein” or "Intact Deconvolution" using the appropriate Sequence Editor.
| No. | Description | No. | Description |
|---|---|---|---|
1 | Enter experiment name | 5 | Select processing method |
2 | Load raw files and assign conditions | 6 | Open the processing method editing wizard |
3 | Specify result format | 7 | Submit experiment to the run queue for automatic processing |
4 | Select protein or oligonucleotide sequence(s) | 8 | Open the Process and Review page to manually process experiment |
Prerequisites
- (Optional) You have added a protein or oligonucleotide sequence to the Sequence Manager.
Procedure
- On the BioPharma Finder Home page, in the Experiment Types pane, select the Intact Mass Analysis workflow link.
- The Intact Mass Analysis page opens.
- In the Intact Mass Analysis Definition area, type the name of the experiment in the Experiment Name box.
TIP
Experiment names can contain up to 50 characters, including alphanumeric, space, underscore (“_”), and period (“.”) characters.
IMPORTANT
You can overwrite a completed experiment by entering the same experiment name.
You cannot overwrite an experiment that is currently submitted to the run queue or is actively processing or canceling in the run queue.
- In the Load Raw Data area, select the Select Raw Data (“...”) button.
- The Add Analysis File(s) dialog box opens.
NOTE
The default file location for raw data files is
C:\Xcalibur\data.- In the Add Analysis File(s) dialog box, locate and select the raw data files (.raw) you want to add to the experiment.
TIP
To add a set of raw data files to the experiment (for example, replicates of the same sample or samples with various experiment conditions), select contiguous files using the SHIFT key or noncontiguous files using the CTRL key.
- Select Open.
- The application adds the imported raw data files to the table in the Load Raw Data area.
- The order of the files in this table defines the order that they appear in the Results tables on the Process and Review page.
- In the Load Raw Data area, in the Condition box, type the experiment condition for each raw data file, separated by a space.
- If an entered condition matches a portion of a raw data file name, the application automatically assigns that condition to one or more matching raw data files.
- To change the condition assigned to a raw data file, use the drop-down menu in the Condition column next to the corresponding raw file name to select a different condition.
- In the Reference Condition box, select the reference condition for the experiment from the drop-down menu.
NOTE
The default reference condition is the first entered condition in the Condition box.
- The application uses the indicated reference condition to calculate various analytical ratios during processing. These ratios are reported in the Results table(s) on the Process and Review page.
- If you load multiple raw data files for an experiment, in the Result Format for Multiple Raw Files area, select either the Batch Processing or Multiconsensus option.
TIP
For more information on these results formats, see Result formats: Batch Processing and Multiconsensus.
NOTE
For Intact Mass Analysis and Top Down Analysis experiments, you can process up to 10 raw data files using the Multiconsensus results format.
IMPORTANT
The Batch Processing results format is not supported for Intact Mass Analysis experiments processed in Manual mode.
You must select the Multiconsensus results format to process an Intact Mass Analysis in Manual mode.
- (Optional) In the Sequence area, select the checkbox in the table row for up to 10 sequences you want to assign to the experiment.
- The application applies the selected sequence to the experiment during processing.
IMPORTANT
Selecting a sequence is required for target sequence matching Intact Mass Analysis experiments.
TIP
If you initially process an experiment without applying a sequence, you can identify components later by importing or creating a sequence and then using real-time optimization (RTO) to apply the sequence to your results.
- In the Processing Method area, select the checkbox for the processing method you want to use or customize for your experiment.
NOTE
IonTrap data acquired in centroid mode is not supported by either the ReSpect or Xtract algorithm.
For IonTrap data acquired in profile mode, you must use a processing method with the ReSpect deconvolution algorithm. The application cannot process IonTrap (profile mode) data with the Xtract deconvolution algorithm.
- (Optional) (Local mode only) Select the Enable Export for External Reporting checkbox to automatically create an intact report workbook (ZIP file) for each batch experiment on the local PC.
NOTE
The automatic intact report workbook generation feature is not available whenconnected to the Ardia Platform.
- (Optional) For Sliding Windows experiments, select the Enable Automatic Sliding Window Parameter Values checkbox if you want the application to automatically determine several parameters for the experiment using the raw data files.
- Then, do one of the following:
- Select Add to Queue to submit your experiment for processing and open the Queue page.
- Select Edit Method to open the processing method editing wizard and edit the experiment parameters before processing.
- Select Manual Process to transfer to the Process and Review page where you can manually process the experiment.