You can run an Intact Mass Analysis experiment in Automatic or Manual mode:
- In Automatic mode, the application automatically processes your experiment using the parameters in the selected processing method.
- In Manual mode, you can determine the optimal settings for selecting chromatographic peaks and deconvolving the spectrum before processing your experiment. After you determine the optimal settings, you can save them to a processing method which will be available for use in Automatic mode for subsequent experiments.
IMPORTANT
The Batch Processing results format is not supported for Intact Mass Analysis experiments processed in Manual mode.
You must select the Multiconsensus results format to process an Intact Mass Analysis in Manual mode.
IMPORTANT
The Auto Peak Detection deconvolution method is not supported for Intact Mass Analysis experiments processed in Manual mode.
You must select a processing method using the Average Over Selected Retention Time or Sliding Windows deconvolution method to process an Intact Mass Analysis in Manual mode.
Manual mode | Automatic mode | |
|---|---|---|
Steps | In Manual mode, you must manually perform the following:
| In Automatic mode, the application automatically performs the following:
|
Results format |
|
|
Deconvolution algorithm |
|
|
Deconvolution methods |
|
|
Action on the Intact Mass Analysis page | Select the Manual Process button to open the Process and Review page and manually specify experiment settings. | Select the Add to Queue button to add the experiment to the run queue for processing. |